missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAND1 Protein
A cDNA sequence encoding the HAND1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12100-1mg
Additional Details : Weight : 0.01000kg
Specifications
HAND1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNLVGSY AHHHHHHHPH PAHPMLHEPF LFGPASRCHQ ERPYFQSWLL SPADAAPDFP AGGPPPAAAA AATAYGPDAR PGQSPGRLEA LGGRLGRRKG SGPKKERRRT ESINSAFAEL RECIPNVPAD TKLSKIKTLR LATSYIAYLM DVLAKDAQSG DPEAFKAELK KADGGRESKR KRELQQHEGF PPALGPVEKR IKGRTGWPQQ VWALELNQ | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HAND1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |