missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HAND1 Protein

A cDNA sequence encoding the HAND1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12100-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949368

  • £155.00 / 20µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HAND1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMNLVGSY AHHHHHHHPH PAHPMLHEPF LFGPASRCHQ ERPYFQSWLL SPADAAPDFP AGGPPPAAAA AATAYGPDAR PGQSPGRLEA LGGRLGRRKG SGPKKERRRT ESINSAFAEL RECIPNVPAD TKLSKIKTLR LATSYIAYLM DVLAKDAQSG DPEAFKAELK KADGGRESKR KRELQQHEGF PPALGPVEKR IKGRTGWPQQ VWALELNQ
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HAND1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers