missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNB5 Protein
A DNA sequence encoding the Homo sapiens (Human) GNB5, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_006569.1 |
---|---|
Gene ID (Entrez) | 10681 |
Name | GNB5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050820
|
enQuireBio™
QP6103-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070820
|
enQuireBio™
QP6103-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060820
|
enQuireBio™
QP6103-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080820
|
enQuireBio™
QP6103-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090820
|
enQuireBio™
QP6103-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000830
|
enQuireBio™
QP6103-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006569.1 | |
GNB5 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10681 | |
Research Use Only | |
GNB5 | |
Recombinant Protein | |
E. coli | |
MATEGLHENE TLASLKSEAE SLKGKLEEER AKLHDVELHQ VAERVEALGQ FVMKTRRTLK GHGNKVLCMD WCKDKRRIVS SSQDGKVIVW DSFTTNKEHA VTMPCTWVMA CAYAPSGCAI ACGGLDNKCS VYPLTFDKNE NMAAKKKSVA MHTNYLSACS FTNSDMQILT ASGDGTCALW DVESGQLLQS FHGHGADVLC LDLAPSETGN TFVSGGCDKK AMVWDMRSGQ CVQAFETHES DINSVRYYPS GDAFASGSDD ATCRLYDLRA DREVAIYSKE SIIFGASSVD FSLSGRLLFA GYNDYTINVW DVLKGSRVSI LFGHENRVST LRVSPDGTAF CSGSWDHTLR VWA | |
Greater than 90% as determined by SDS-PAGE. |