missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNB5 Protein

A DNA sequence encoding the Homo sapiens (Human) GNB5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6103-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070820

  • £1735.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006569.1
GNB5 Protein
Research Use Only
GNB5
Recombinant Protein
E. coli
MATEGLHENE TLASLKSEAE SLKGKLEEER AKLHDVELHQ VAERVEALGQ FVMKTRRTLK GHGNKVLCMD WCKDKRRIVS SSQDGKVIVW DSFTTNKEHA VTMPCTWVMA CAYAPSGCAI ACGGLDNKCS VYPLTFDKNE NMAAKKKSVA MHTNYLSACS FTNSDMQILT ASGDGTCALW DVESGQLLQS FHGHGADVLC LDLAPSETGN TFVSGGCDKK AMVWDMRSGQ CVQAFETHES DINSVRYYPS GDAFASGSDD ATCRLYDLRA DREVAIYSKE SIIFGASSVD FSLSGRLLFA GYNDYTINVW DVLKGSRVSI LFGHENRVST LRVSPDGTAF CSGSWDHTLR VWA
Greater than 90% as determined by SDS-PAGE.
10681
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers