missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glia-derived nexin Protein
A DNA sequence encoding the Homo sapiens (Human) Glia-derived nexin, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001130000.1 |
---|---|
Gene ID (Entrez) | 5270 |
Name | Glia-derived nexin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065000
|
enQuireBio™
QP6674-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16085000
|
enQuireBio™
QP6674-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075000
|
enQuireBio™
QP6674-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095000
|
enQuireBio™
QP6674-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005010
|
enQuireBio™
QP6674-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015010
|
enQuireBio™
QP6674-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001130000.1 | |
Glia-derived nexin Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5270 | |
Research Use Only | |
SERPINE2 | |
Recombinant Protein | |
E. coli | |
SHFNPLSLEE LGSNTGIQVF NQIVKSRPHD NIVISPHGIA SVLGMLQLGA DGRTKKQLAM VMRYGVNGVG KILKKINKAI VSKKNKDIVT VANAVFVKNA SEIEVPFVTR NKDVFQCEVR NVNFEDPASA CDSINAWVKN ETRDMIDNLL SPDLIDGVLT RLVLVNAVYF KGLWKSRFQP ENTKKRTFVA ADGKSYQVPM LAQLSVFRCG STSAPNDLWY NFIELPYHGE SISMLIALPT ESSTPLSAII PHISTKTIDS WMSIMVPKRV QVILPKFTAV AQTDLKEPLK VLGITDMFDS SKANFAKITT GSENLHVSHI LQKAKIEVSE DGTKASAATT AILIARSSPP WFIVDRPFLF FIRHNPTGAV LFMGQINKP | |
Greater than 90% as determined by SDS-PAGE. |