missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glia-derived nexin Protein
A DNA sequence encoding the Homo sapiens (Human) Glia-derived nexin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6674-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001130000.1 | |
Glia-derived nexin Protein | |
Research Use Only | |
SERPINE2 | |
Recombinant Protein | |
E. coli | |
SHFNPLSLEE LGSNTGIQVF NQIVKSRPHD NIVISPHGIA SVLGMLQLGA DGRTKKQLAM VMRYGVNGVG KILKKINKAI VSKKNKDIVT VANAVFVKNA SEIEVPFVTR NKDVFQCEVR NVNFEDPASA CDSINAWVKN ETRDMIDNLL SPDLIDGVLT RLVLVNAVYF KGLWKSRFQP ENTKKRTFVA ADGKSYQVPM LAQLSVFRCG STSAPNDLWY NFIELPYHGE SISMLIALPT ESSTPLSAII PHISTKTIDS WMSIMVPKRV QVILPKFTAV AQTDLKEPLK VLGITDMFDS SKANFAKITT GSENLHVSHI LQKAKIEVSE DGTKASAATT AILIARSSPP WFIVDRPFLF FIRHNPTGAV LFMGQINKP | |
Greater than 90% as determined by SDS-PAGE. |
5270 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |