missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF7 Protein
A cDNA sequence encoding the GDF7 was constructed and used to recombinantly synthesize the protein.
£104.00 - £5425.00
Specifications
Name | GDF7 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | Untagged |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974458
|
enQuireBio™
QP10628-2UG |
2 μg |
£104.00
2µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15954458
|
enQuireBio™
QP10628-10UG |
10 μg |
£159.00
10µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15964458
|
enQuireBio™
QP10628-1MG |
1 mg |
£5425.00
1mg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
GDF7 Protein | |
Untagged | |
Human | |
Greater than 95.0% as determined by SDS-PAGE. | |
GDF7 |
Recombinant Protein | |
E. coli | |
TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR | |
151449 |