missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF7 Protein
A cDNA sequence encoding the GDF7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10628-10ug
Additional Details : Weight : 0.01000kg
Specifications
151449 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50, as determined by inducing alkaline phosphatase production by mouse ATDC5 cells, is less than 1.25microg/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in HCl. |
GDF7 Protein | |
Research Use Only | |
GDF7 | |
Recombinant Protein | |
E. coli | |
TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR | |
Greater than 95.0% as determined by SDS-PAGE. |