missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF7 Protein

A cDNA sequence encoding the GDF7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10628-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15974458

  • £99.15 / 2µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

151449
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50, as determined by inducing alkaline phosphatase production by mouse ATDC5 cells, is less than 1.25microg/ml.
Human
Untagged
Lyophilized from a 0.2microm filtered solution in HCl.
GDF7 Protein
Research Use Only
GDF7
Recombinant Protein
E. coli
TALAGTRTAQ GSGGGAGRGH GRRGRSRCSR KPLHVDFKEL GWDDWIIAPL DYEAYHCEGL CDFPLRSHLE PTNHAIIQTL LNSMAPDAAP ASCCVPARLS PISILYIDAA NNVVYKQYED MVVEACGCR
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers