missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibrinogen beta chain Protein
A DNA sequence encoding the Homo sapiens (Human) Fibrinogen beta chain, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001171670.1 |
---|---|
Gene ID (Entrez) | 2244 |
Name | Fibrinogen beta chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919519
|
enQuireBio™
QP6035-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939519
|
enQuireBio™
QP6035-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929519
|
enQuireBio™
QP6035-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949519
|
enQuireBio™
QP6035-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959519
|
enQuireBio™
QP6035-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969519
|
enQuireBio™
QP6035-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001171670.1 | |
Fibrinogen beta chain Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2244 | |
Research Use Only | |
FGB | |
Recombinant Protein | |
E. coli | |
GHRPLDKKRE EAPSLRPAPP PISGGGYRAR PAKAAATQKK VERKAPDAGG CLHADPDLGV LCPTGCQLQE ALLQQERPIR NSVDELNNNV EAVSQTSSSS FQYMYLLKDL WQKRQKQVKD NENVVNEYSS ELEKHQLYID ETVNSNIPTN LRVLRSILEN LRSKIQKLES DVSAQMEYCR TPCTVSCNIP VVSGKECEEI IRKGGETSEM YLIQPDSSVK PYRVYCDMNT ENGGWTVIQN RQDGSVDFGR KWDPYKQGFG NVATNTDGKN YCGLPGEYWL GNDKISQLTR MGPTELLIEM EDWKGDKVKA HYGGFTVQNE ANKYQISVNK YRGTAGNALM DGASQLMGEN RTMTIHNGMF FSTYDRDNDG WLTSDPRKQC SKEDGGGWWY NRCHAANPNG RYYWGGQYTW DMAKHGTDDG VVWMNWKGSW YSMRKMSMKI RPFFPQQ | |
Greater than 90% as determined by SDS-PAGE. |