missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibrinogen beta chain Protein
A DNA sequence encoding the Homo sapiens (Human) Fibrinogen beta chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6035-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171670.1 | |
Fibrinogen beta chain Protein | |
Research Use Only | |
FGB | |
Recombinant Protein | |
E. coli | |
GHRPLDKKRE EAPSLRPAPP PISGGGYRAR PAKAAATQKK VERKAPDAGG CLHADPDLGV LCPTGCQLQE ALLQQERPIR NSVDELNNNV EAVSQTSSSS FQYMYLLKDL WQKRQKQVKD NENVVNEYSS ELEKHQLYID ETVNSNIPTN LRVLRSILEN LRSKIQKLES DVSAQMEYCR TPCTVSCNIP VVSGKECEEI IRKGGETSEM YLIQPDSSVK PYRVYCDMNT ENGGWTVIQN RQDGSVDFGR KWDPYKQGFG NVATNTDGKN YCGLPGEYWL GNDKISQLTR MGPTELLIEM EDWKGDKVKA HYGGFTVQNE ANKYQISVNK YRGTAGNALM DGASQLMGEN RTMTIHNGMF FSTYDRDNDG WLTSDPRKQC SKEDGGGWWY NRCHAANPNG RYYWGGQYTW DMAKHGTDDG VVWMNWKGSW YSMRKMSMKI RPFFPQQ | |
Greater than 90% as determined by SDS-PAGE. |
2244 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |