missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF10 / KGF2 Protein
A DNA sequence encoding the Human FGF10 / KGF2 Protein, was expressed in the hosts and tags indicated.
£95.54 - £1797.47
Specifications
Accession Number | NP_004456.1 |
---|---|
Gene ID (Entrez) | 2255 |
Name | FGF10 / KGF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934558
|
enQuireBio™
QP1066-EC-5UG |
5 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15924558
|
enQuireBio™
QP1066-EC-25UG |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15904558
|
enQuireBio™
QP1066-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15914558
|
enQuireBio™
QP1066-EC-250UG |
250 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004456.1 | |
FGF10 / KGF2 Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LGQDMVSPEA TNSSSSSFSS PSSAGRHVRS YNHLQGDVRW RKLFSFTKYF LKIEKNGKVS GTKKENCPYS ILEITSVEIG VVAVKAINSN YYLAMNKKGK LYGSKEFNND CKLKERIEEN GYNTYASFNW QHNGRQMYVA LNGKGAPRRG QKTRRKNTSA HFLPMVVHS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
2255 | |
Research Use Only | |
FGF10 | |
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 2 M PBS, pH 7.4, lyophilized |