missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF10 / KGF2 Protein
A DNA sequence encoding the Human FGF10 / KGF2 Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP1066-ec-250ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004456.1 | |
FGF10 / KGF2 Protein | |
Research Use Only | |
FGF10 | |
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 2 M PBS, pH 7.4, lyophilized |
2255 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LGQDMVSPEA TNSSSSSFSS PSSAGRHVRS YNHLQGDVRW RKLFSFTKYF LKIEKNGKVS GTKKENCPYS ILEITSVEIG VVAVKAINSN YYLAMNKKGK LYGSKEFNND CKLKERIEEN GYNTYASFNW QHNGRQMYVA LNGKGAPRRG QKTRRKNTSA HFLPMVVHS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |