missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF2B2 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF2B2, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_055054.1 |
---|---|
Gene ID (Entrez) | 8892 |
Name | EIF2B2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929029
|
enQuireBio™
QP5970-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949029
|
enQuireBio™
QP5970-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939029
|
enQuireBio™
QP5970-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959029
|
enQuireBio™
QP5970-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969029
|
enQuireBio™
QP5970-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979029
|
enQuireBio™
QP5970-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055054.1 | |
EIF2B2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
8892 | |
Research Use Only | |
EIF2B2 | |
Recombinant Protein | |
E. coli | |
MPGSAAKGSE LSERIESFVE TLKRGGGPRS SEEMARETLG LLRQIITDHR WSNAGELMEL IRREGRRMTA AQPSETTVGN MVRRVLKIIR EEYGRLHGRS DESDQQESLH KLLTSGGLNE DFSFHYAQLQ SNIIEAINEL LVELEGTMEN IAAQALEHIH SNEVIMTIGF SRTVEAFLKE AARKRKFHVI VAECAPFCQG HEMAVNLSKA GIETTVMTDA AIFAVMSRVN KVIIGTKTIL ANGALRAVTG THTLALAAKH HSTPLIVCAP MFKLSPQFPN EEDSFHKFVA PEEVLPFTEG DILEKVSVHC PVFDYVPPEL ITLFISNIGG NAPSYIYRLM SELYHPDDHV L | |
Greater than 80% as determined by SDS-PAGE. |