missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF2B2 Protein

A DNA sequence encoding the Homo sapiens (Human) EIF2B2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5970-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949029

  • £2480.00 / 1mg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055054.1
EIF2B2 Protein
Research Use Only
EIF2B2
Recombinant Protein
E. coli
MPGSAAKGSE LSERIESFVE TLKRGGGPRS SEEMARETLG LLRQIITDHR WSNAGELMEL IRREGRRMTA AQPSETTVGN MVRRVLKIIR EEYGRLHGRS DESDQQESLH KLLTSGGLNE DFSFHYAQLQ SNIIEAINEL LVELEGTMEN IAAQALEHIH SNEVIMTIGF SRTVEAFLKE AARKRKFHVI VAECAPFCQG HEMAVNLSKA GIETTVMTDA AIFAVMSRVN KVIIGTKTIL ANGALRAVTG THTLALAAKH HSTPLIVCAP MFKLSPQFPN EEDSFHKFVA PEEVLPFTEG DILEKVSVHC PVFDYVPPEL ITLFISNIGG NAPSYIYRLM SELYHPDDHV L
Greater than 80% as determined by SDS-PAGE.
8892
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers