missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTP synthase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) CTP synthase 1, was expressed in the hosts and tags indicated.
£183.90 - £2010.00
Specifications
Accession Number | NP_001288166.1 |
---|---|
Gene ID (Entrez) | 1503 |
Name | CTP synthase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047611
|
enQuireBio™
QP9088-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067611
|
enQuireBio™
QP9088-YE-1MG |
1 mg |
£2010.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057611
|
enQuireBio™
QP9088-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077611
|
enQuireBio™
QP9088-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087611
|
enQuireBio™
QP9088-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097611
|
enQuireBio™
QP9088-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001288166.1 | |
CTP synthase 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1503 | |
Research Use Only | |
CTPS1 | |
Recombinant Protein | |
Yeast | |
MKYILVTGGV ISGIGKGIIA SSVGTILKSC GLHVTSIKID PYINIDAGTF SPYEHGEVFV LDDGGEVDLD LGNYERFLDI RLTKDNNLTT GKIYQYVINK ERKGDYLGKT VQVVPHITDA IQEWVMRQAL IPVDEDGLEP QVCVIELGGT VGDIESMPFI EAFRQFQFKV KRENFCNIHV SLVPQPSSTG EQKTKPTQNS VRELRGLGLS PDLVVCRCSN PLDTSVKEKI SMFCHVEPEQ VICVHDVSSI YRVPLLLEEQ GVVDYFLRRL DLPIERQPRK MLMKWKEMAD RYDRLLETCS IALVGKYTKF SDSYASVIKA LEHSALAINH KLEIKYIDSA DLEPITSQEE PVRYHEAWQK LCSAHGVLVP GGFGVRGTEG KIQAIAWARN QKKPFLGVCL GMQLAVVEFS RNVLGWQDAN STEFDPTTSH PVVVDMPEHN PGQMGGTMRL GKRRTLFQTK NSVMRKLYGD ADYLEERHRH RFEVNPVWKK CLEEQGLKFV GQDVEGERME IVELEDHPFF VGVQYHPEFL SRPIKPSPPY FGLLLASVGR LSHYLQKGCR LSPRDTYSDR SGSSSPDSEI TELKFPSINH D | |
Greater than 90% as determined by SDS-PAGE. |