missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTP synthase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) CTP synthase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9088-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001288166.1 | |
CTP synthase 1 Protein | |
Research Use Only | |
CTPS1 | |
Recombinant Protein | |
Yeast | |
MKYILVTGGV ISGIGKGIIA SSVGTILKSC GLHVTSIKID PYINIDAGTF SPYEHGEVFV LDDGGEVDLD LGNYERFLDI RLTKDNNLTT GKIYQYVINK ERKGDYLGKT VQVVPHITDA IQEWVMRQAL IPVDEDGLEP QVCVIELGGT VGDIESMPFI EAFRQFQFKV KRENFCNIHV SLVPQPSSTG EQKTKPTQNS VRELRGLGLS PDLVVCRCSN PLDTSVKEKI SMFCHVEPEQ VICVHDVSSI YRVPLLLEEQ GVVDYFLRRL DLPIERQPRK MLMKWKEMAD RYDRLLETCS IALVGKYTKF SDSYASVIKA LEHSALAINH KLEIKYIDSA DLEPITSQEE PVRYHEAWQK LCSAHGVLVP GGFGVRGTEG KIQAIAWARN QKKPFLGVCL GMQLAVVEFS RNVLGWQDAN STEFDPTTSH PVVVDMPEHN PGQMGGTMRL GKRRTLFQTK NSVMRKLYGD ADYLEERHRH RFEVNPVWKK CLEEQGLKFV GQDVEGERME IVELEDHPFF VGVQYHPEFL SRPIKPSPPY FGLLLASVGR LSHYLQKGCR LSPRDTYSDR SGSSSPDSEI TELKFPSINH D | |
Greater than 90% as determined by SDS-PAGE. |
1503 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |