missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDKN2C Protein
A cDNA sequence encoding the CDKN2C was constructed and used to recombinantly synthesize the protein.
£104.00 - £3130.00
Specifications
Name | CDKN2C Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947078
|
enQuireBio™
QP11378-5UG |
5 μg |
£104.00
5µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15937078
|
enQuireBio™
QP11378-20UG |
20 μg |
£159.00
20µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15927078
|
enQuireBio™
QP11378-1MG |
1 mg |
£3130.00
1mg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
CDKN2C Protein | |
His | |
Human | |
Greater than 95% as determined by SDS-PAGE. | |
CDKN2C |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAEPWG NELASAAARG DLEQLTSLLQ NNVNVNAQNG FGRTALQVMK LGNPEIARRL LLRGANPDLK DRTGFAVIHD AARAGFLDTL QTLLEFQADV NIEDNEGNLP LHLAAKEGHL RVVEFLVKHT ASNVGHRNHK GDTACDLARL YGRNEVVSLM QANGAGGATN LQ | |
1031 |