missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDKN2C Protein
A cDNA sequence encoding the CDKN2C was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11378-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
1031 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAEPWG NELASAAARG DLEQLTSLLQ NNVNVNAQNG FGRTALQVMK LGNPEIARRL LLRGANPDLK DRTGFAVIHD AARAGFLDTL QTLLEFQADV NIEDNEGNLP LHLAAKEGHL RVVEFLVKHT ASNVGHRNHK GDTACDLARL YGRNEVVSLM QANGAGGATN LQ | |
Greater than 95% as determined by SDS-PAGE. |
CDKN2C Protein | |
Research Use Only | |
CDKN2C | |
Human | |
His | |
The CDKN2C solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 10% glycerol. |