missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDKN2C Protein

A cDNA sequence encoding the CDKN2C was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11378-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947078

  • £98.39 / 5µg
Estimated Shipment: 01-03-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1031
Greater than 95% as determined by SDS-PAGE.
Research Use Only
CDKN2C
Human
His
The CDKN2C solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 10% glycerol.
CDKN2C Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAEPWG NELASAAARG DLEQLTSLLQ NNVNVNAQNG FGRTALQVMK LGNPEIARRL LLRGANPDLK DRTGFAVIHD AARAGFLDTL QTLLEFQADV NIEDNEGNLP LHLAAKEGHL RVVEFLVKHT ASNVGHRNHK GDTACDLARL YGRNEVVSLM QANGAGGATN LQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers