missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD19 / Leu-12 Protein
A DNA sequence encoding the Recombinant Homo sapiens (Human) CD19 / Leu-12, was expressed in the hosts and tags indicated.
£155.00 - £2655.00
Specifications
Accession Number | NP_001171569.1 |
---|---|
Gene ID (Entrez) | 930 |
Name | CD19 / Leu-12 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967709
|
enQuireBio™
QP5782-EC-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907719
|
enQuireBio™
QP5782-EC-50UG |
50 μg |
£329.00
50µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957709
|
enQuireBio™
QP5782-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987709
|
enQuireBio™
QP5782-EC-200UG |
200 μg |
£965.00
200µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997709
|
enQuireBio™
QP5782-EC-500UG |
500 μg |
£1645.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977709
|
enQuireBio™
QP5782-EC-1MG |
1 mg |
£2655.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001171569.1 | |
CD19 / Leu-12 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
930 | |
Research Use Only | |
CD19 | |
Recombinant Protein | |
E. coli | |
PEEPLVVKVE EGDNAVLQCL KGTSDGPTQQ LTWSRESPLK PFLKLSLGLP GLGIHMRPLA IWLFIFNVSQ QMGGFYLCQP GPPSEKAWQP GWTVNVEGSG ELFRWNVSDL GGLGCGLKNR SSEGPSSPSG KLMSPKLYVW AKDRPEIWEG EPPCLPPRDS LNQSLSQDLT MAPGSTLWLS CGVPPDSVSR GPLSWTHVHP KGPKSLLSLE LKDDRPARDM WVMETGLLLP RATAQDAGKY YCHRGNLTMS FHLEITARPV LWHWLLRTGG WK | |
Greater than 90% as determined by SDS-PAGE. |