missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD19 / Leu-12 Protein

A DNA sequence encoding the Recombinant Homo sapiens (Human) CD19 / Leu-12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5782-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987709

  • £965.00 / 200µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001171569.1
CD19 / Leu-12 Protein
Research Use Only
CD19
Recombinant Protein
E. coli
PEEPLVVKVE EGDNAVLQCL KGTSDGPTQQ LTWSRESPLK PFLKLSLGLP GLGIHMRPLA IWLFIFNVSQ QMGGFYLCQP GPPSEKAWQP GWTVNVEGSG ELFRWNVSDL GGLGCGLKNR SSEGPSSPSG KLMSPKLYVW AKDRPEIWEG EPPCLPPRDS LNQSLSQDLT MAPGSTLWLS CGVPPDSVSR GPLSWTHVHP KGPKSLLSLE LKDDRPARDM WVMETGLLLP RATAQDAGKY YCHRGNLTMS FHLEITARPV LWHWLLRTGG WK
Greater than 90% as determined by SDS-PAGE.
930
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers