missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD19 / Leu-12 Protein
A DNA sequence encoding the Recombinant Homo sapiens (Human) CD19 / Leu-12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5782-ec-50ug
Additional Details : Weight : 0.00900kg
Specifications
NP_001171569.1 | |
CD19 / Leu-12 Protein | |
Research Use Only | |
CD19 | |
Recombinant Protein | |
E. coli | |
PEEPLVVKVE EGDNAVLQCL KGTSDGPTQQ LTWSRESPLK PFLKLSLGLP GLGIHMRPLA IWLFIFNVSQ QMGGFYLCQP GPPSEKAWQP GWTVNVEGSG ELFRWNVSDL GGLGCGLKNR SSEGPSSPSG KLMSPKLYVW AKDRPEIWEG EPPCLPPRDS LNQSLSQDLT MAPGSTLWLS CGVPPDSVSR GPLSWTHVHP KGPKSLLSLE LKDDRPARDM WVMETGLLLP RATAQDAGKY YCHRGNLTMS FHLEITARPV LWHWLLRTGG WK | |
Greater than 90% as determined by SDS-PAGE. |
930 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |