missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRAK Protein
A cDNA sequence encoding the BRAK was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 9547 |
---|---|
Name | BRAK Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | CXCL14 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944178
|
enQuireBio™
QP10538-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934178
|
enQuireBio™
QP10538-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924178
|
enQuireBio™
QP10538-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
9547 | |
Research Use Only | |
CXCL14 | |
Recombinant Protein | |
E. coli | |
MSLLPRRAPP VSMRLLAAAL LLLLLALYTA RVDGSKCKCS RKGPKIRYSD VKKLEMKPKY PHCEEKMVII TTKSVSRYRG QEHCLHPKLQ STKRFIKWYN AWNEKRRVYE E | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
BRAK Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 of CXCL14 as determined by its ability to induce calcium flux of prostaglandin E2 treated THP1 human acute monocytic leukemia cells was 1.0-10.0 ng/ml. | |
Human | |
Untagged | |
CXCL14 was lyophilized after extensive dialysis against 20mM Tris-HCl, pH 8.5 and 1M NaCl. |