missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BRAK Protein

A cDNA sequence encoding the BRAK was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10538-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944178

  • £99.15 / 5µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

9547
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 of CXCL14 as determined by its ability to induce calcium flux of prostaglandin E2 treated THP1 human acute monocytic leukemia cells was 1.0-10.0 ng/ml.
Human
Untagged
CXCL14 was lyophilized after extensive dialysis against 20mM Tris-HCl, pH 8.5 and 1M NaCl.
BRAK Protein
Research Use Only
CXCL14
Recombinant Protein
E. coli
MSLLPRRAPP VSMRLLAAAL LLLLLALYTA RVDGSKCKCS RKGPKIRYSD VKKLEMKPKY PHCEEKMVII TTKSVSRYRG QEHCLHPKLQ STKRFIKWYN AWNEKRRVYE E
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers