missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRAK Protein
A cDNA sequence encoding the BRAK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10538-1mg
Additional Details : Weight : 0.01000kg
Specifications
9547 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 of CXCL14 as determined by its ability to induce calcium flux of prostaglandin E2 treated THP1 human acute monocytic leukemia cells was 1.0-10.0 ng/ml. | |
Human | |
Untagged | |
CXCL14 was lyophilized after extensive dialysis against 20mM Tris-HCl, pH 8.5 and 1M NaCl. |
BRAK Protein | |
Research Use Only | |
CXCL14 | |
Recombinant Protein | |
E. coli | |
MSLLPRRAPP VSMRLLAAAL LLLLLALYTA RVDGSKCKCS RKGPKIRYSD VKKLEMKPKY PHCEEKMVII TTKSVSRYRG QEHCLHPKLQ STKRFIKWYN AWNEKRRVYE E | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |