missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AIF1L Protein
A cDNA sequence encoding the AIF1L was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 83543 |
---|---|
Name | AIF1L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | AIF1L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945788
|
enQuireBio™
QP10987-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935788
|
enQuireBio™
QP10987-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925788
|
enQuireBio™
QP10987-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
83543 | |
Research Use Only | |
AIF1L | |
Human | |
His | |
The AIF1L protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1mM NaCl and 20% glycerol. |
AIF1L Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGELSNRFQ GGKAFGLLKA RQERRLAEIN REFLCDQKYS DEENLPEKLT AFKEKYMEFD LNNEGEIDLM SLKRMMEKLG VPKTHLEMKK MISEVTGGVS DTISYRDFVN MMLGKRSAVL KLVMMFEGKA NESSPKPVGP PPERDIASLP | |
Greater than 95% as determined by SDS-PAGE. |