missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AIF1L Protein
A cDNA sequence encoding the AIF1L was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10987-20ug
Additional Details : Weight : 0.01000kg
Specifications
83543 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGELSNRFQ GGKAFGLLKA RQERRLAEIN REFLCDQKYS DEENLPEKLT AFKEKYMEFD LNNEGEIDLM SLKRMMEKLG VPKTHLEMKK MISEVTGGVS DTISYRDFVN MMLGKRSAVL KLVMMFEGKA NESSPKPVGP PPERDIASLP | |
Greater than 95% as determined by SDS-PAGE. |
AIF1L Protein | |
Research Use Only | |
AIF1L | |
Human | |
His | |
The AIF1L protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1mM NaCl and 20% glycerol. |