missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AIF1L Protein

A cDNA sequence encoding the AIF1L was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10987-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945788

  • £98.39 / 5µg
Estimated Shipment: 18-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

83543
Greater than 95% as determined by SDS-PAGE.
Research Use Only
AIF1L
Human
His
The AIF1L protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1mM NaCl and 20% glycerol.
AIF1L Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSGELSNRFQ GGKAFGLLKA RQERRLAEIN REFLCDQKYS DEENLPEKLT AFKEKYMEFD LNNEGEIDLM SLKRMMEKLG VPKTHLEMKK MISEVTGGVS DTISYRDFVN MMLGKRSAVL KLVMMFEGKA NESSPKPVGP PPERDIASLP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers