missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster Glutamine synthetase Protein
A DNA sequence encoding the Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus) Glutamine synthetase, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_001233699.1 |
---|---|
Gene ID (Entrez) | 100689337 |
Name | Glutamine synthetase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050790
|
enQuireBio™
QP6099-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070790
|
enQuireBio™
QP6099-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060790
|
enQuireBio™
QP6099-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080790
|
enQuireBio™
QP6099-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090790
|
enQuireBio™
QP6099-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000800
|
enQuireBio™
QP6099-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001233699.1 | |
Glutamine synthetase Protein | |
Not determined. | |
Full Length | |
Chinese Hamster | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
100689337 | |
Research Use Only | |
GLUL | |
Recombinant Protein | |
E. coli | |
ATSASSHLNK GIKQMYMSLP QGEKVQAMYI WVDGTGEGLR CKTRTLDCEP KCVEELPEWN FDGSSTFQSE SSNSDMYLSP VAMFRDPFRK EPNKLVFCEV FKYNQKPAET NLRHTCKRIM DMVSNQHPWF GMEQEYTLLG TDGHPFGWPS DGFPGPQGLY YCGVGADKAY RRDIMEAHYR ACLYAGVKIT GTYAEVKHAQ WEFQIGPCEG IRMGDHLWVA RFILHRVCKD FGVIATFDSK PIPGNWNGAG CHTNFSTKTM REENGLKHIK EAIEKLSKRH RYHIRAYDPK GGLDNARRLT GFHKTSNIND FSAGVADRSA SIRIPRTVGQ EKKGYFEARC PSANCDPFAV TEAIVRTCLL NETGDQPFQY KN | |
Greater than 90% as determined by SDS-PAGE. |