missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster Glutamine synthetase Protein
A DNA sequence encoding the Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus) Glutamine synthetase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6099-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001233699.1 | |
Glutamine synthetase Protein | |
Research Use Only | |
GLUL | |
Recombinant Protein | |
E. coli | |
ATSASSHLNK GIKQMYMSLP QGEKVQAMYI WVDGTGEGLR CKTRTLDCEP KCVEELPEWN FDGSSTFQSE SSNSDMYLSP VAMFRDPFRK EPNKLVFCEV FKYNQKPAET NLRHTCKRIM DMVSNQHPWF GMEQEYTLLG TDGHPFGWPS DGFPGPQGLY YCGVGADKAY RRDIMEAHYR ACLYAGVKIT GTYAEVKHAQ WEFQIGPCEG IRMGDHLWVA RFILHRVCKD FGVIATFDSK PIPGNWNGAG CHTNFSTKTM REENGLKHIK EAIEKLSKRH RYHIRAYDPK GGLDNARRLT GFHKTSNIND FSAGVADRSA SIRIPRTVGQ EKKGYFEARC PSANCDPFAV TEAIVRTCLL NETGDQPFQY KN | |
Greater than 90% as determined by SDS-PAGE. |
100689337 | |
500 μg | |
Not determined. | |
Full Length | |
Chinese Hamster | |
His-SUMO | |
Tris-based buffer, 50% glycerol |