missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine PTX3 / Pentraxin 3 / TSG-14 Protein
A DNA sequence encoding the Bos taurus (Bovine) PTX3 / Pentraxin 3 / TSG-14, was expressed in the hosts and tags indicated.
£463.00 - £1295.00
Specifications
Name | PTX3 / Pentraxin 3 / TSG-14 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | Flag |
Host Species | Mammal |
Cross Reactivity | Bovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962878
|
enQuireBio™
QP10121-MA-20UG |
20 μg |
£463.00
20µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15952878
|
enQuireBio™
QP10121-MA-100UG |
100 μg |
£1295.00
100µg |
Estimated Shipment: 20-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15972878
|
enQuireBio™
QP10121-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Specifications
PTX3 / Pentraxin 3 / TSG-14 Protein | |
Flag | |
Bovine | |
Greater than 90% as determined by SDS-PAGE. | |
NP_001069727.1 |
Recombinant Protein | |
Mammal | |
ENSDDYELMY VNLDNEIDNG LHPTEDPTPC DCSRENSEWD KLFTMLENSQ MREGMLLQAT DVMLRGELQK LQAELGRLEG SLQKLCGPEA PSETRLARAL DDLLQASRDA GRRLARLEDA GALRPQEEAG RALGAVLEEL RRTRADLRAV QGWAASRWLP AGCETAILFP MRSKKIFASV HPVTPMKLET FSACIWVKAT EVLNKTVLFS YGTKRNPYEI QLYLSYRSIM LVVGGEENRL VADAVISLGT WTHLCSTWDS KKGHMALWVN GDSVATAVDM ATGHVVPEGG ILQIGQEKNG CCVGGGFDET LAFSGRLTGF NIWEGVLSNE EIREAGGAES CHIRGNVVGW GVTEIQPHGG AQYVY | |
541148 | |
PTX3 |