missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine PTX3 / Pentraxin 3 / TSG-14 Protein

A DNA sequence encoding the Bos taurus (Bovine) PTX3 / Pentraxin 3 / TSG-14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10121-ma-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962878

  • £439.00 / 20µg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001069727.1
PTX3 / Pentraxin 3 / TSG-14 Protein
Research Use Only
PTX3
Recombinant Protein
Mammal
ENSDDYELMY VNLDNEIDNG LHPTEDPTPC DCSRENSEWD KLFTMLENSQ MREGMLLQAT DVMLRGELQK LQAELGRLEG SLQKLCGPEA PSETRLARAL DDLLQASRDA GRRLARLEDA GALRPQEEAG RALGAVLEEL RRTRADLRAV QGWAASRWLP AGCETAILFP MRSKKIFASV HPVTPMKLET FSACIWVKAT EVLNKTVLFS YGTKRNPYEI QLYLSYRSIM LVVGGEENRL VADAVISLGT WTHLCSTWDS KKGHMALWVN GDSVATAVDM ATGHVVPEGG ILQIGQEKNG CCVGGGFDET LAFSGRLTGF NIWEGVLSNE EIREAGGAES CHIRGNVVGW GVTEIQPHGG AQYVY
Greater than 90% as determined by SDS-PAGE.
541148
20 μg
Not determined.
Full Length
Bovine
Flag
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers