missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine PTX3 / Pentraxin 3 / TSG-14 Protein
A DNA sequence encoding the Bos taurus (Bovine) PTX3 / Pentraxin 3 / TSG-14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10121-ma-100ug
This item is not returnable.
View return policy
Specifications
NP_001069727.1 | |
PTX3 / Pentraxin 3 / TSG-14 Protein | |
Research Use Only | |
PTX3 | |
Recombinant Protein | |
Mammal | |
ENSDDYELMY VNLDNEIDNG LHPTEDPTPC DCSRENSEWD KLFTMLENSQ MREGMLLQAT DVMLRGELQK LQAELGRLEG SLQKLCGPEA PSETRLARAL DDLLQASRDA GRRLARLEDA GALRPQEEAG RALGAVLEEL RRTRADLRAV QGWAASRWLP AGCETAILFP MRSKKIFASV HPVTPMKLET FSACIWVKAT EVLNKTVLFS YGTKRNPYEI QLYLSYRSIM LVVGGEENRL VADAVISLGT WTHLCSTWDS KKGHMALWVN GDSVATAVDM ATGHVVPEGG ILQIGQEKNG CCVGGGFDET LAFSGRLTGF NIWEGVLSNE EIREAGGAES CHIRGNVVGW GVTEIQPHGG AQYVY | |
Greater than 90% as determined by SDS-PAGE. |
541148 | |
100 μg | |
Not determined. | |
Full Length | |
Bovine | |
Flag | |
Tris-based buffer, 50% glycerol |