missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. baumannii Outer membrane protein Omp38 Protein
A DNA sequence encoding the Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377) Outer membrane protein Omp38, was expressed in the hosts and tags indicated.
£321.05 - £2888.42
Specifications
Name | Outer membrane protein Omp38 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | omp38 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068800
|
enQuireBio™
QP7424-YE-100UG |
100 μg |
£800.00
100µg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008810
|
enQuireBio™
QP7424-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078800
|
enQuireBio™
QP7424-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098800
|
enQuireBio™
QP7424-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018810
|
enQuireBio™
QP7424-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088800
|
enQuireBio™
QP7424-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Outer membrane protein Omp38 Protein | |
Not determined. | |
Full Length | |
A. baumannii | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
omp38 | |
Recombinant Protein | |
Yeast | |
ANAGVTVTPL LLGYTFQDSQ HNNGGKDGNL TNGPELQDDL FVGAALGIEL TPWLGFEAEY NQVKGDVDGA SAGAEYKQKQ INGNFYVTSD LITKNYDSKI KPYVLLGAGH YKYDFDGVNR GTRGTSEEGT LGNAGVGAFW RLNDALSLRT EARATYNADE EFWNYTALAG LNVVLGGHLK PAAPVVEVAP VEPTPVTPQP QELTEDLNME LRVFFDTNKS NIKDQYKPEI AKVAEKLSEY PNATARIEGH TDNTGPRKLN ERLSLARANS VKSALVNEYN VDASRLSTQG FAWDQPIADN KTKEGRAMNR RVFATITGSR TVVVQPGQEA AAPAAAQ | |
Greater than 90% as determined by SDS-PAGE. |