missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant A. baumannii Outer membrane protein Omp38 Protein

A DNA sequence encoding the Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377) Outer membrane protein Omp38, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7424-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068800

  • £800.00 / 100µg
Estimated Shipment: 24-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Outer membrane protein Omp38 Protein
Research Use Only
omp38
Recombinant Protein
Yeast
ANAGVTVTPL LLGYTFQDSQ HNNGGKDGNL TNGPELQDDL FVGAALGIEL TPWLGFEAEY NQVKGDVDGA SAGAEYKQKQ INGNFYVTSD LITKNYDSKI KPYVLLGAGH YKYDFDGVNR GTRGTSEEGT LGNAGVGAFW RLNDALSLRT EARATYNADE EFWNYTALAG LNVVLGGHLK PAAPVVEVAP VEPTPVTPQP QELTEDLNME LRVFFDTNKS NIKDQYKPEI AKVAEKLSEY PNATARIEGH TDNTGPRKLN ERLSLARANS VKSALVNEYN VDASRLSTQG FAWDQPIADN KTKEGRAMNR RVFATITGSR TVVVQPGQEA AAPAAAQ
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
A. baumannii
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers