missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. baumannii Beta-lactamase Protein
A DNA sequence encoding the Acinetobacter baumannii Beta-lactamase, was expressed in the hosts and tags indicated.
£281.57 - £2410.48
Specifications
Accession Number | WP_004201164.1 |
---|---|
Gene ID (Entrez) | 14971909 |
Name | Beta-lactamase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065401
|
enQuireBio™
QP8581-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005411
|
enQuireBio™
QP8581-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055401
|
enQuireBio™
QP8581-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085401
|
enQuireBio™
QP8581-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095401
|
enQuireBio™
QP8581-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075401
|
enQuireBio™
QP8581-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_004201164.1 | |
Beta-lactamase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
A. baumannii | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
14971909 | |
Research Use Only | |
NDM-1 | |
Recombinant Protein | |
E. coli | |
AANGWVEPAT APNFGPLKVF YPGPGHTSDN ITVGIDGTDI AFGGCLIKDS KAKSLGNLG | |
Greater than 90% as determined by SDS-PAGE. |