missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. baumannii Beta-lactamase Protein
A DNA sequence encoding the Acinetobacter baumannii Beta-lactamase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8581-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
WP_004201164.1 | |
Beta-lactamase Protein | |
Research Use Only | |
NDM-1 | |
Recombinant Protein | |
E. coli | |
AANGWVEPAT APNFGPLKVF YPGPGHTSDN ITVGIDGTDI AFGGCLIKDS KAKSLGNLG | |
Greater than 90% as determined by SDS-PAGE. |
14971909 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
A. baumannii | |
His-SUMO | |
Tris-based buffer, 50% glycerol |