missing translation for 'onlineSavingsMsg'
Learn More

Mouse IL-23R Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 9 publications

£168.00 - £1530.00

Specifications

Antigen IL-23R
Clone 258010
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL
Applications Western Blot, Neutralization
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18763427
GREENER_CHOICE
View Documents
R&D Systems
MAB1686-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18782057
GREENER_CHOICE
View Documents
R&D Systems
MAB1686-100
100 μg
£395.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18781197
GREENER_CHOICE
View Documents
R&D Systems
MAB1686-500
500 μg
£1530.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications

Specifications

IL-23R
LYOPH
Western Blot, Neutralization
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
IL23R
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
258010
Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL
Monoclonal
Rat
Mouse
Q8K4B4
149233
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession # Q8K4B4
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse IL-23 R in Western blots. In Western blots, this antibody does not cross-react with recombinant human IL-23 R.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.