Learn More
Mouse IL-23R Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 9 publications
Brand:  R&D Systems MAB1686-SP
 View more versions of this product
Product Code. 18763427
  • £168.00 / 25µg
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-23R
Monoclonal
LYOPH
Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL
Q8K4B4
IL23R
Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession # Q8K4B4
25 μg
Primary
Detects mouse IL-23 R in Western blots. In Western blots, this antibody does not cross-react with recombinant human IL-23 R.
Mouse
Purified
Western Blot, Neutralization
258010
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
149233
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b