missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-23R Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB1686-500
This item is not returnable.
View return policy
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-23R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| Q8K4B4 | |
| IL23R | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R Gly24-Asp372 Accession # Q8K4B4 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-23 R in Western blots. In Western blots, this antibody does not cross-react with recombinant human IL-23 R. | |
| Mouse | |
| Supernatant |
| Western Blot, Neutralization | |
| 258010 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 149233 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction