missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | Rad17 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 3-5ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18199212
|
Novus Biologicals
NBP2-27106SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18114535
|
Novus Biologicals
NBP2-27106 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rad17 Polyclonal specifically detects Rad17 in Human, Mouse, Primate samples. It is validated for Western Blot.Specifications
| Rad17 | |
| Western Blot 3-5ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
| RAD17 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_f and CRA_d), rhesus monkey, chimpanzee and orangutan, 92% homologous in horse, cow and mouse and 85% homologous in dog and rat. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Primate | |
| O75943 | |
| 5884 | |
| This antibody was raised against RAD17 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title