missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27106
This item is not returnable.
View return policy
Description
Rad17 Polyclonal specifically detects Rad17 in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications
| Rad17 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_f and CRA_d), rhesus monkey, chimpanzee and orangutan, 92% homologous in horse, cow and mouse and 85% homologous in dog and rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 3-5ug/ml | |
| O75943 | |
| RAD17 | |
| This antibody was raised against RAD17 | |
| 0.1 mg | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
| 5884 | |
| Human, Mouse, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction