missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27106SS
This item is not returnable.
View return policy
Description
Rad17 Polyclonal specifically detects Rad17 in Human, Mouse, Primate samples. It is validated for Western Blot.
Specifications
| Rad17 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_f and CRA_d), rhesus monkey, chimpanzee and orangutan, 92% homologous in horse, cow and mouse and 85% homologous in dog and rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 3-5ug/ml | |
| O75943 | |
| RAD17 | |
| This antibody was raised against RAD17 | |
| 0.025 mg | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
| 5884 | |
| Human, Mouse, Primate | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido