missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB3C Recombinant Rabbit Monoclonal Antibody (10X10S0)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen RAB3C
Clone 10X10S0
Concentration 1.18 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283898
View Documents
Invitrogen™
MA557987
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282947
GREENER_CHOICE
View Documents
Invitrogen™
MA557988
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: IKTYSWDNAQ VILVGNKCDM EDERVISTER GQHLGEQLGF EFFETSAKDN INVKQTFERL VDIICDKMSE SLETDPAITA AKQNTRLKET PPPPQPNCAC.

Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 3C is found in brain, adrenal gland, and pituitary.
Specifications

Specifications

RAB3C
1.18 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P62823, P62824, Q96E17
115827, 171058, 67295
Synthetic peptide.
Antibody
10X10S0
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
RAB3C
2700062I01Rik; 3110015B08Rik; 3110037E15Rik; AI850886; RAB3C; RAB3C, member RAS oncogene family; ras-related protein Rab-3C; small GTP-binding protein Rab3C
RAB3C
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.