missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB3C Recombinant Rabbit Monoclonal Antibody (10X10S0)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557988

 View more versions of this product

Product Code. 30282947

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IKTYSWDNAQ VILVGNKCDM EDERVISTER GQHLGEQLGF EFFETSAKDN INVKQTFERL VDIICDKMSE SLETDPAITA AKQNTRLKET PPPPQPNCAC.

Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 3C is found in brain, adrenal gland, and pituitary.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAB3C
Recombinant Monoclonal
1.18 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P62823, P62824, Q96E17
RAB3C
Synthetic peptide.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
10X10S0
Unconjugated
RAB3C
2700062I01Rik; 3110015B08Rik; 3110037E15Rik; AI850886; RAB3C; RAB3C, member RAS oncogene family; ras-related protein Rab-3C; small GTP-binding protein Rab3C
Rabbit
Protein A
RUO
115827, 171058, 67295
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado