missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB3C Recombinant Rabbit Monoclonal Antibody (10X10S0)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557987

 View more versions of this product

Product Code. 30283898

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IKTYSWDNAQ VILVGNKCDM EDERVISTER GQHLGEQLGF EFFETSAKDN INVKQTFERL VDIICDKMSE SLETDPAITA AKQNTRLKET PPPPQPNCAC.

Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 3C is found in brain, adrenal gland, and pituitary.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAB3C
Recombinant Monoclonal
1.18 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P62823, P62824, Q96E17
RAB3C
Synthetic peptide.
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
10X10S0
Unconjugated
RAB3C
2700062I01Rik; 3110015B08Rik; 3110037E15Rik; AI850886; RAB3C; RAB3C, member RAS oncogene family; ras-related protein Rab-3C; small GTP-binding protein Rab3C
Rabbit
Protein A
RUO
115827, 171058, 67295
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.