missing translation for 'onlineSavingsMsg'
Learn More

Quinapril hydrochloride, 98%, Thermo Scientific™

Angiotensin-converting enzyme inhibitor

£31.40 - £445.02

Chemical Identifiers

CAS 82586-55-8
Molecular Formula C25H31ClN2O5
Molecular Weight (g/mol) 474.982
MDL Number MFCD00889215
InChI Key IBBLRJGOOANPTQ-JKVLGAQCSA-N
Synonym quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
PubChem CID 54891
ChEBI CHEBI:8714
IUPAC Name (3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
SMILES CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494399
SDS View Documents
Thermo Scientific Alfa Aesar
J61913.MC
100mg
ONLINE EXCLUSIVE PRICE
£32.10
£31.40 / 100mg
Save £0.70
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15474399
SDS View Documents
Thermo Scientific Alfa Aesar
J61913.03
1g
ONLINE EXCLUSIVE PRICE
£181.00
£177.00 / 1g
Save £4.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15484399
SDS View Documents
Thermo Scientific Alfa Aesar
J61913.06
5g
ONLINE EXCLUSIVE PRICE
£455.00
£445.02 / 5g
Save £9.98
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

82586-55-8
474.982
IBBLRJGOOANPTQ-JKVLGAQCSA-N
54891
(3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
C25H31ClN2O5
MFCD00889215
quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
CHEBI:8714
CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl

Specifications

82586-55-8
MFCD00889215
quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl
474.982
CHEBI:8714
98%
C25H31ClN2O5
14,8051
IBBLRJGOOANPTQ-JKVLGAQCSA-N
(3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
54891
474.99
Quinapril hydrochloride
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361

missing translation for 'rtecsNumber' : NW7176000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.