missing translation for 'onlineSavingsMsg'
Learn More

Quinapril hydrochloride, 98%, Thermo Scientific™

Angiotensin-converting enzyme inhibitor

Brand:  Thermo Scientific Alfa Aesar J61913.03

Quantity 1g

 View more versions of this product

Product Code. 15474399

  • ONLINE EXCLUSIVE PRICE
    £181.00
    £177.00 / 1g
    Save £4.00
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

82586-55-8
474.982
IBBLRJGOOANPTQ-JKVLGAQCSA-N
54891
(3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
C25H31ClN2O5
MFCD00889215
quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
CHEBI:8714
CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl

Specifications

82586-55-8
MFCD00889215
14,8051
IBBLRJGOOANPTQ-JKVLGAQCSA-N
(3S)-2-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid;hydrochloride
54891
474.99
Quinapril hydrochloride
C25H31ClN2O5
1g
quinapril hydrochloride, accupril, accuprin, acequin, quinazil, korec, quinapril hcl, lidaltrin, acuitel, ectren
CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CC3=CC=CC=C3CC2C(=O)O.Cl
474.982
CHEBI:8714
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.