missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ 60mm Diameter Glass Büchner Filter Funnel

Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.

Brand:  Pyrex 3700/10

Code : NEW

Additional Details : Weight : 0.66000kg

 View more versions of this product

Product Code. 15567710

  • £790.00 / Pack of 10
Estimated Shipment: 27-09-2024
to see stock.
Add to basket

Description

Description

Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 60mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.

  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
  • Choice of disc diameters and porosities
Specifications

Specifications

125 mL
Pyrex Glass, Borosilicate Glass
For convenient filtration of solutions
Integrated filter plate sintered Glass disc
5
13mm
180 mm
60mm
100mm
1.0 to 1.6μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers